accessibility menu, dialog, popup

UserName

CAS RN 209664-72-2

NOOOONH2K

CAS RN 209664-72-2

IUPAC Name: potassium 2-amino-5-nitrobenzoate
Synonyms: potassium 5-nitroanthranilate
Molecular Weight (g/mol): 220.23
Molecular Formula: C7H5KN2O4
InChi Key: VUOPWTVKTDYMCB-UHFFFAOYSA-M
SMILES: [K+].NC1=C(C=C(C=C1)[N+]([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results