accessibility menu, dialog, popup

UserName

CAS RN 2096336-51-3

BSOONHHOOH

CAS RN 2096336-51-3

IUPAC Name: [3-(phenylsulfamoyl)phenyl]boronic acid
Synonyms: 3-(phenylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 277.1
Molecular Formula: C12H12BNO4S
InChi Key: SPCPUZDVDUTVFG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)S(=O)(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results