accessibility menu, dialog, popup

UserName

FNCSF

CAS RN 206559-57-1

IUPAC Name: 1,4-difluoro-2-isothiocyanatobenzene
Synonyms: 1,4-difluoro-2-isothiocyanatobenzene
Molecular Weight (g/mol): 171.16
Molecular Formula: C7H3F2NS
InChi Key: QVIDXDRFMDPVLA-UHFFFAOYSA-N
SMILES: FC1=CC(N=C=S)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results