CAS RN 2037-95-8
CAS RN 2037-95-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Carsalam, MedChemExpress
MedChemExpress Carsalam (Carbonylsalicylamide) is a nonsteroidal anti-inflammatory drug.