CAS RN 20323-74-4
CAS RN 20323-74-4
IUPAC Name:
ethyl 2-amino-4,5-dimethoxybenzoate
Synonyms:
ethyl 2-amino-4,5-dimethoxybenzoate
Molecular Weight (g/mol):
225.24
Molecular Formula:
C11H15NO4
InChi Key:
SMICMEHDDWELMR-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(OC)=C(OC)C=C1N
1
–
1
of
1
results
1
Ethyl 2-amino-4,5-dimethoxybenzoate, 98%, Thermo Scientific™
CAS: 20323-74-4 Molecular Formula: C11H15NO4 Molecular Weight (g/mol): 225.244 MDL Number: MFCD00017323 InChI Key: SMICMEHDDWELMR-UHFFFAOYSA-N Synonym: ethyl 6-amino-3,4-dimethoxybenzoate,benzoic acid, 2-amino-4,5-dimethoxy-, ethyl ester,6-amino-3,4-dimethoxybenzoic acid ethyl ester,2-amino-4,5-dimethoxybenzoic acid ethyl ester,ethyl 4,5-dimethoxyanthranilate,ethyl2-amino-4,5-dimethoxybenzoate,acmc-20a83g PubChem CID: 88488 IUPAC Name: ethyl 2-amino-4,5-dimethoxybenzoate SMILES: CCOC(=O)C1=CC(=C(C=C1N)OC)OC