accessibility menu, dialog, popup

UserName

CH3IBr

CAS RN 202865-85-8

IUPAC Name: 1-bromo-4-iodo-2-methylbenzene
Synonyms: 1-bromo-4-iodo-2-methylbenzene
Molecular Weight (g/mol): 296.93
Molecular Formula: C7H6BrI
InChi Key: QSQOGKONVJDRNH-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results