CAS RN 199329-53-8
CAS RN 199329-53-8
IUPAC Name:
disodium 2-amino-3-[(2-amino-2-carboxylatoethyl)disulfanyl]propanoate hydrate
Synonyms:
disodium 2-amino-3-[(2-amino-2-carboxylatoethyl)disulfanyl]propanoate hydrate
Molecular Weight (g/mol):
302.27
Molecular Formula:
C6H12N2Na2O5S2
InChi Key:
TVPDKJXPEMKMDM-UHFFFAOYNA-L
SMILES:
O.[Na+].[Na+].NC(CSSCC(N)C([O-])=O)C([O-])=O
1
–
2
of
2
results
1
L-Cystine disodium salt monohydrate, 98+%
CAS: 199329-53-8 Molecular Formula: C6H12N2Na2O5S2 Molecular Weight (g/mol): 302.271 MDL Number: MFCD00243291 InChI Key: TVPDKJXPEMKMDM-BLZBDRIDSA-L Synonym: disodium l-cystine anion hydrate,l-cystine disodium salt monohydrate,h-cys-oh 2.2na.h2o,c6h10n2o4s2.2na.h2o PubChem CID: 71299670 IUPAC Name: disodium;(2R)-2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate;hydrate SMILES: C(C(C(=O)[O-])N)SSCC(C(=O)[O-])N.O.[Na+].[Na+]
2
Melford L-Cystine Disodium Salt Monohydrate
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.