accessibility menu, dialog, popup

UserName

CAS RN 19781-77-2

H3COHCH2

CAS RN 19781-77-2

IUPAC Name: hept-6-en-3-ol
Synonyms: hept-6-en-3-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: XFXWEAWJVWCOBF-UHFFFAOYNA-N
SMILES: CCC(O)CCC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results