accessibility menu, dialog, popup

UserName

CAS RN 19293-60-8

H3CNH2N

CAS RN 19293-60-8

IUPAC Name: 1-(1-methyl-1H-indol-3-yl)methanamine
Synonyms: 1-(1-methylindol-3-yl)methanamine
Molecular Weight (g/mol): 160.22
Molecular Formula: C10H12N2
InChi Key: NOFZMDGMQKRLIV-UHFFFAOYSA-N
SMILES: CN1C=C(CN)C2=CC=CC=C12
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)