accessibility menu, dialog, popup

UserName

CAS RN 19009-39-3

H3CCH3NCH3CH3ClO

CAS RN 19009-39-3

IUPAC Name: N,N-bis(propan-2-yl)carbamoyl chloride
Synonyms: N,N-diisopropylcarbamoyl chloride
Molecular Weight (g/mol): 163.65
Molecular Formula: C7H14ClNO
InChi Key: RSAFAYLZKCYUQW-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)