accessibility menu, dialog, popup

UserName

CAS RN 190004-53-6

OHONHOO

CAS RN 190004-53-6

IUPAC Name: 1-{[(benzyloxy)carbonyl]amino}cyclobutane-1-carboxylic acid
Synonyms: 1-{[(benzyloxy)carbonyl]amino}cyclobutane-1-carboxylic acid
Molecular Weight (g/mol): 249.27
Molecular Formula: C13H15NO4
InChi Key: GDJSFBNRXFOUEQ-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCC1)NC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)