CAS RN 189808-70-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress NH2-PEG3-C1-Boc
MedChemExpress NH2-PEG3-C1-Boc (PROTAC Linker 5) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs.