accessibility menu, dialog, popup

UserName

ClNCSCl

CAS RN 18967-42-5

IUPAC Name: 1,2-dichloro-4-(isothiocyanatomethyl)benzene
Synonyms: 1,2-dichloro-4-(isothiocyanatomethyl)benzene
Molecular Weight (g/mol): 218.1
Molecular Formula: C8H5Cl2NS
InChi Key: DPSSHXWTNIGVMQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CN=C=S)C=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results