accessibility menu, dialog, popup

UserName

CAS RN 189628-39-5

CH3OF

CAS RN 189628-39-5

IUPAC Name: 3-fluoro-5-methylbenzaldehyde
Synonyms: 3-fluoro-5-methylbenzaldehyde
Molecular Weight (g/mol): 138.14
Molecular Formula: C8H7FO
InChi Key: XXPITUUQKCGWNR-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results