CAS RN 18656-96-7
CAS RN 18656-96-7
IUPAC Name:
cyclohexanaminium 6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Synonyms:
cyclohexylammonium ion 6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular Weight (g/mol):
521.79
Molecular Formula:
C20H26BrClN2O7
InChi Key:
JXCKZXHCJOVIAV-UHFFFAOYNA-N
SMILES:
[NH3+]C1CCCCC1.OC1C(O)C(OC2=CNC3=CC=C(Br)C(Cl)=C23)OC(C1O)C([O-])=O
1
–
2
of
2
results
1
MedChemExpress X-Gluc Dicyclohexylamine
MedChemExpress X-Gluc Dicyclohexylamine is used as a reagent to detect β-glucuronidase, an enzyme produced by the E.
2
MedChemExpress X-Gluc Dicyclohexylamine
MedChemExpress X-Gluc Dicyclohexylamine is used as a reagent to detect β-glucuronidase, an enzyme produced by the E.