CAS RN 18404-43-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Digitoxigenin Mono-Beta-D-digitoxoside, TRC
CAS: 18404-43-8 Molecular Formula: C29H44O7 Molecular Weight (g/mol): 504.66 Synonym: 3β-[(2,6-Dideoxy-β-D-ribo-hexopyranosyl)oxy]-14-hydroxy-5β-card-20(22)-enolide,Evatromonoside,(3β,5β)-3-[(2,6-Dideoxy-β-D-ribo-hexopyranosyl)oxy]-14-hydroxycard-20(22)-enolide,2,6-Dideoxy-β-D-ribo-hexopyranoside-digitoxigenin-3,2,6-Dideoxy-β-D-ribo-hexopyranoside Digitoxigenin,Digitoxigenin Digitoxoside Digitoxigenin Monodigitoxoside SMILES: C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]2CC[C@@]3(C)[C@H](CC[C@@H]4[C@@H]3CC[C@]5(C)[C@H](CC[C@]45O)C6=CC(=O)OC6)C2