CAS RN 182563-08-2
CAS RN 182563-08-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
LY334370, MedChemExpress
MedChemExpress LY334370 is a selective 5-HT1F receptor agonist with a Ki of 1.6 nM.