accessibility menu, dialog, popup

UserName

CAS RN 181217-39-0

NHClHOHOHOHCl

CAS RN 181217-39-0

IUPAC Name: 6-chloro-1-(4-hydroxyphenyl)-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol hydrochloride
Synonyms: fenoldopam hydrochloride
Molecular Weight (g/mol): 342.22
Molecular Formula: C16H17Cl2NO3
InChi Key: NLMPGIXLXSPNFS-UHFFFAOYNA-N
SMILES: Cl.OC1=CC=C(C=C1)C1CNCCC2=C(Cl)C(O)=C(O)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results