accessibility menu, dialog, popup

UserName

CAS RN 180981-09-3

NOHNOOHNHONHOHNONONONNH2H2NNHONNH2H2NNH2OHOHNNNH2H2NOOH

CAS RN 180981-09-3

IUPAC Name: 2-({1-[2-(2-{2-[2-({1-[1-(2-{2-amino-5-[(diaminomethylidene)amino]pentanamido}-5-[(diaminomethylidene)amino]pentanoyl)pyrrolidine-2-carbonyl]-4-hydroxypyrrolidin-2-yl}formamido)acetamido]-2-(2,3-dihydro-1H-inden-2-yl)acetamido}-3-hydroxypropanamido)-2-(2,3-dihydro-1H-inden-2-yl)acetyl]-octahydro-1H-indol-2-yl}formamido)-5-[(diaminomethylidene)amino]pentanoic acid
Synonyms: 2-({1-[2-(2-{2-[2-({1-[1-(2-{2-amino-5-[(diaminomethylidene)amino]pentanamido}-5-[(diaminomethylidene)amino]pentanoyl)pyrrolidine-2-carbonyl]-4-hydroxypyrrolidin-2-yl}formamido)acetamido]-2-(2,3-dihydro-1H-inden-2-yl)acetamido}-3-hydroxypropanamido)-2-(2,3-dihydro-1H-inden-2-yl)acetyl]-octahydroindol-2-yl}formamido)-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Weight (g/mol): 1338.58
Molecular Formula: C64H95N19O13
InChi Key: PVNQRMOWAXFQHU-UHFFFAOYNA-N
SMILES: NC(CCCN=C(N)N)C(=O)NC(CCCN=C(N)N)C(=O)N1CCCC1C(=O)N1CC(O)CC1C(=O)NCC(=O)NC(C1CC2=CC=CC=C2C1)C(=O)NC(CO)C(=O)NC(C1CC2=CC=CC=C2C1)C(=O)N1C2CCCCC2CC1C(=O)NC(CCCN=C(N)N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)