CAS RN 18063-02-0
CAS RN 18063-02-0
IUPAC Name:
2,6-difluorobenzoyl chloride
Synonyms:
2,6-difluorobenzoyl chloride
Molecular Weight (g/mol):
176.55
Molecular Formula:
C7H3ClF2O
InChi Key:
QRHUZEVERIHEPT-UHFFFAOYSA-N
SMILES:
FC1=CC=CC(F)=C1C(Cl)=O
1
–
2
of
2
results
1
2,6-Difluorobenzoyl chloride, 98%
CAS: 18063-02-0 Molecular Formula: C7H3ClF2O Molecular Weight (g/mol): 176.547 MDL Number: MFCD00000659 InChI Key: QRHUZEVERIHEPT-UHFFFAOYSA-N Synonym: 2,6-difluorobenzoylchloride,benzoyl chloride, 2,6-difluoro,2,6-difluoro-benzoyl chloride,acmc-1ca8w,2,6-diflurobenzoyl chloride,2 6-difluorobenzoyl chloride,2,6-diflourobenzoyl chloride,2,6-difluoro benzoylchloride,2.6-difluorobenzoyl chloride,2,6-difluro-benzoyl chloride PubChem CID: 87438 IUPAC Name: 2,6-difluorobenzoyl chloride SMILES: C1=CC(=C(C(=C1)F)C(=O)Cl)F