accessibility menu, dialog, popup

UserName

SNOOH

CAS RN 179408-54-9

IUPAC Name: 6-(thiophen-2-yl)pyridine-3-carboxylic acid
Synonyms: 6-(thiophen-2-yl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 205.23
Molecular Formula: C10H7NO2S
InChi Key: QYGFXNUFKLRCRB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(C=C1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results