CAS RN 177952-39-5
CAS RN 177952-39-5
IUPAC Name:
2-[2,4-bis(trifluoromethyl)phenyl]acetic acid
Synonyms:
[2,4-bis(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol):
272.15
Molecular Formula:
C10H6F6O2
InChi Key:
BCHGLBXVODTIEE-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC=C(C=C1C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
2,4-Bis(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™
CAS: 177952-39-5 Molecular Formula: C10H6F6O2 Molecular Weight (g/mol): 272.15 MDL Number: MFCD00042492 InChI Key: BCHGLBXVODTIEE-UHFFFAOYSA-N Synonym: 2,4-bis trifluoromethyl phenylacetic acid,2,4-ditrifluoromethylphenylacetic acid,2-2,4-bis trifluoromethyl phenyl acetic acid,2,4-bis trifluoromethyl phenyl acetic acid,rarechem al bo 0404,2,4-bis-trifluoromethyl-phenyl-acetic acid,benzeneacetic acid, 2,4-bis trifluoromethyl,2,4-bis trifluoromethyl-phenyl acetic acid,pubchem2199,acmc-209xc8 PubChem CID: 2736172 IUPAC Name: 2-[2,4-bis(trifluoromethyl)phenyl]acetic acid SMILES: OC(=O)CC1=CC=C(C=C1C(F)(F)F)C(F)(F)F