CAS RN 17605-06-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl 3-Oxohept-6-enoate, TRC
CAS: 17605-06-0 Molecular Formula: C9 H14 O3 Molecular Weight (g/mol): 170.21 IUPAC Name: ethyl 3-oxohept-6-enoate SMILES: CCOC(=O)CC(=O)CCC=C