accessibility menu, dialog, popup

UserName

CAS RN 17572-09-7

OSNH2

CAS RN 17572-09-7

IUPAC Name: furan-2-carbothioamide
Synonyms: furan-2-carbothioamide
Molecular Weight (g/mol): 127.16
Molecular Formula: C5H5NOS
InChi Key: OUZNHXMJEBIDSZ-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results