accessibility menu, dialog, popup

UserName

CAS RN 175291-56-2

H3C(R)O(S)HNOHOPOOOHOO

CAS RN 175291-56-2

IUPAC Name: (2R,3S)-3-{[(benzyloxy)(hydroxy)phosphoryl]oxy}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (2R,3S)-3-{[benzyloxy(hydroxy)phosphoryl]oxy}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 511.47
Molecular Formula: C26H26NO8P
InChi Key: HOFDVXHILSPFNS-BXKMTCNYSA-N
SMILES: C[C@H](OP(O)(=O)OCC1=CC=CC=C1)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)