accessibility menu, dialog, popup

UserName

CAS RN 173952-44-8

H3CNHONNNH2OOCH3

CAS RN 173952-44-8

IUPAC Name: 8-(4-aminophenyl)-5-methyl-N-propyl-2H,5H,6H-[1,3]dioxolo[4,5-g]phthalazine-6-carboxamide
Synonyms: 8-(4-aminophenyl)-5-methyl-N-propyl-2H,5H-[1,3]dioxolo[4,5-g]phthalazine-6-carboxamide
Molecular Weight (g/mol): 366.42
Molecular Formula: C20H22N4O3
InChi Key: OFUDZKKOKPGXOH-UHFFFAOYNA-N
SMILES: CCCNC(=O)N1N=C(C2=CC=C(N)C=C2)C2=CC3=C(OCO3)C=C2C1C
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results