CAS RN 17071-54-4
IUPAC Name:
1-(hexyloxy)octane
Synonyms:
1-(hexyloxy)octane
Molecular Weight (g/mol):
214.39
Molecular Formula:
C14H30O
InChi Key:
KUTWKGLRMXBROO-UHFFFAOYSA-N
SMILES:
CCCCCCCCOCCCCCC
1
–
1
of
1
results
1
n-Hexyl n-octyl ether, 98%, Thermo Scientific™
CAS: 17071-54-4 Molecular Formula: C14H30O Molecular Weight (g/mol): 214.393 MDL Number: MFCD00049132 InChI Key: KUTWKGLRMXBROO-UHFFFAOYSA-N Synonym: hexyl octyl ether,n-hexyl n-octyl ether,1-hexyloxy octane,octylhexyl ether,1-n-hexyloxyoctane,1-hexyloxy octane # PubChem CID: 519362 IUPAC Name: 1-hexoxyoctane SMILES: CCCCCCCCOCCCCCC