accessibility menu, dialog, popup

UserName

CH3OH3CSOOCl

CAS RN 166964-26-7

IUPAC Name: 2,5-dimethylfuran-3-sulfonyl chloride
Synonyms: 2,5-dimethylfuran-3-sulfonyl chloride
Molecular Weight (g/mol): 194.63
Molecular Formula: C6H7ClO3S
InChi Key: PIXAQZAVANAGLE-UHFFFAOYSA-N
SMILES: CC1=CC(=C(C)O1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results