accessibility menu, dialog, popup

UserName

CH3NNNOOOHNOONNSOONH3CH3CH3C(?)

CAS RN 1668553-26-1

IUPAC Name: 7-[5-({4-[4-(dimethylsulfamoyl)piperazin-1-yl]phenoxy}methyl)-1,3-dimethyl-1H-pyrazol-4-yl]-1-[2-(morpholin-4-yl)ethyl]-3-[3-(naphthalen-1-yloxy)propyl]-1H-indole-2-carboxylic acid
Synonyms: 7-(5-{4-[4-(dimethylsulfamoyl)piperazin-1-yl]phenoxymethyl}-1,3-dimethylpyrazol-4-yl)-1-[2-(morpholin-4-yl)ethyl]-3-[3-(naphthalen-1-yloxy)propyl]indole-2-carboxylic acid
Molecular Weight (g/mol): 850.05
Molecular Formula: C46H55N7O7S
InChi Key: XMVAWGSQPHFXKU-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)N1CCN(CC1)C1=CC=C(OCC2=C(C(C)=NN2C)C2=C3N(CCN4CCOCC4)C(C(O)=O)=C(CCCOC4=C5C=CC=CC5=CC=C4)C3=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)
  • (4)
  • (1)