CAS RN 16577-52-9
IUPAC Name:
lithium(1+) octanoate
Synonyms:
lithium(1+) octanoate
Molecular Weight (g/mol):
150.15
Molecular Formula:
C8H15LiO2
InChi Key:
BTAUEIDLAAYHSL-UHFFFAOYSA-M
SMILES:
[Li+].CCCCCCCC([O-])=O
1
–
1
of
1
results
1
Lithium octanoate, 99%
CAS: 16577-52-9 Molecular Formula: C8H15LiO2 Molecular Weight (g/mol): 150.146 MDL Number: MFCD00210629 InChI Key: BTAUEIDLAAYHSL-UHFFFAOYSA-M Synonym: lithium octanoate,unii-9nq46xlp8b,octanoic acid, lithium salt,9nq46xlp8b,octanoic acid, lithiumsalt 1:1,lithium caprylate,octanoic acid, lithium salt 1:1,acmc-20akmi,lithium 1+ octanoate,lithium 1+ ion octanoate PubChem CID: 23673717 IUPAC Name: lithium;octanoate SMILES: [Li+].CCCCCCCC(=O)[O-]