accessibility menu, dialog, popup

UserName

H3CSOO(S)OOHNHOO

CAS RN 163437-14-7

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methanesulfonylbutanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-methanesulfonylbutanoic acid
Molecular Weight (g/mol): 403.45
Molecular Formula: C20H21NO6S
InChi Key: KJLKPACOHZKRFM-UHFFFAOYNA-N
SMILES: CS(=O)(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)