CAS RN 1626359-59-8
CAS RN 1626359-59-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
sulfo-SPDB-DM4, MedChemExpress
MedChemExpress sulfo-SPDB-DM4 is a drug-linker conjugate for ADC by using the maytansinebased payload (DM4, an antitubulin agent) via the sulfo-SPDB linker.