accessibility menu, dialog, popup

UserName

CAS RN 161957-59-1

H3COClF

CAS RN 161957-59-1

IUPAC Name: 1-(3-chloro-2-fluorophenyl)ethan-1-one
Synonyms: 1-(3-chloro-2-fluorophenyl)ethanone
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: ZHDLNEIXOUIAQO-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C(Cl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results