CAS RN 161928-86-5
CAS RN 161928-86-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
MedChemExpress GP(33-41)
MedChemExpress GP(33-41), a 9-aa-long peptide, is the optimal sequence of the GP1 epitope of lymphocytic choriomeningitis virus, and can upregulate H-2Db molecules at the RMA-S (Db Kb) cell surface with a SC50 of 344 nM.
2
MedChemExpress GP(33-41)
MedChemExpress GP(33-41), a 9-aa-long peptide, is the optimal sequence of the GP1 epitope of lymphocytic choriomeningitis virus, and can upregulate H-2Db molecules at the RMA-S (Db Kb) cell surface with a SC50 of 344 nM.
3
MedChemExpress GP(33-41)
MedChemExpress GP(33-41), a 9-aa-long peptide, is the optimal sequence of the GP1 epitope of lymphocytic choriomeningitis virus, and can upregulate H-2Db molecules at the RMA-S (Db Kb) cell surface with a SC50 of 344 nM.