accessibility menu, dialog, popup

UserName

CAS RN 161446-90-8

HOFCl

CAS RN 161446-90-8

IUPAC Name: (3-chloro-4-fluorophenyl)methanol
Synonyms: (3-chloro-4-fluorophenyl)methanol
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: KRQHBUXUXOPOON-UHFFFAOYSA-N
SMILES: OCC1=CC=C(F)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results