CAS RN 16115-68-7
CAS RN 16115-68-7
IUPAC Name:
1,4-diethyl 2-aminobutanedioate hydrochloride
Synonyms:
1,4-diethyl 2-aminobutanedioate hydrochloride
Molecular Weight (g/mol):
225.67
Molecular Formula:
C8H16ClNO4
InChi Key:
AJOXZAAREAYBQR-UHFFFAOYNA-N
SMILES:
Cl.CCOC(=O)CC(N)C(=O)OCC
1
–
1
of
1
results
1
L-Aspartic acid diethyl ester hydrochloride, 98%
CAS: 16115-68-7 Molecular Formula: C8H16ClNO4 Molecular Weight (g/mol): 225.669 MDL Number: MFCD00038910 InChI Key: AJOXZAAREAYBQR-RGMNGODLSA-N Synonym: l-aspartic acid diethyl ester hydrochloride,h-asp oet-oet.hcl,h-asp oet-oet hcl,s-diethyl 2-aminosuccinate hydrochloride,aspartic acid, diethyl ester, hydrochloride,h-asp oet-oet hydrochloride,diethyl l-aspartate hydrochloride,1,4-diethyl 2s-2-aminobutanedioate hydrochloride,h-asp oet-oet?cl,diethylaspartate hydrochloride PubChem CID: 12416466 IUPAC Name: diethyl (2S)-2-aminobutanedioate;hydrochloride SMILES: CCOC(=O)CC(C(=O)OCC)N.Cl