accessibility menu, dialog, popup

UserName

CAS RN 1609-66-1

CH3ONNH2

CAS RN 1609-66-1

IUPAC Name: 4-(N-phenylpropanamido)piperidin-1-ium
Synonyms: 4-(N-phenylpropanamido)piperidin-1-ium
Molecular Weight (g/mol): 233.33
Molecular Formula: C14H21N2O
InChi Key: PMCBDBWCQQBSRJ-UHFFFAOYSA-O
SMILES: CCC(=O)N(C1CC[NH2+]CC1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results