CAS RN 1607261-66-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Potassium 4-Hydroxy-4-methylpentanoate, TRC
CAS: 1607261-66-4 Molecular Formula: C6H11KO3 Molecular Weight (g/mol): 170.24 IUPAC Name: potassium;4-hydroxy-4-methylpentanoate SMILES: [K+].CC(C)(O)CCC(=O)[O-]