accessibility menu, dialog, popup

UserName

CAS RN 16019-34-4

NNNCl

CAS RN 16019-34-4

IUPAC Name: 7-benzyl-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Synonyms: 7-benzyl-4-chloropyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol): 243.69
Molecular Formula: C13H10ClN3
InChi Key: KMWWCJXJJNHTQL-UHFFFAOYSA-N
SMILES: ClC1=C2C=CN(CC3=CC=CC=C3)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results