accessibility menu, dialog, popup

UserName

CAS RN 160098-96-4

H2NNNNNNNO

CAS RN 160098-96-4

IUPAC Name: 4-(furan-2-yl)-10-(2-phenylethyl)-3,5,6,8,10,11-hexaazatricyclo[7.3.0.0²,⁶]dodeca-1(9),2,4,7,11-pentaen-7-amine
Synonyms: 4-(furan-2-yl)-10-(2-phenylethyl)-3,5,6,8,10,11-hexaazatricyclo[7.3.0.0²,⁶]dodeca-1(9),2,4,7,11-pentaen-7-amine
Molecular Weight (g/mol): 345.37
Molecular Formula: C18H15N7O
InChi Key: UTLPKQYUXOEJIL-UHFFFAOYSA-N
SMILES: NC1=NC2=C(C=NN2CCC2=CC=CC=C2)C2=NC(=NN12)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results