CAS RN 159519-65-0
CAS RN 159519-65-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MedChemExpress Enfuvirtide
MedChemExpress Enfuvirtide (T20;DP178) is an anti-HIV-1 fusion inhibitor peptide.
2
MedChemExpress Enfuvirtide
MedChemExpress Enfuvirtide (T20;DP178) is an anti-HIV-1 fusion inhibitor peptide.