accessibility menu, dialog, popup

UserName

CAS RN 159191-44-3

BNHOOH

CAS RN 159191-44-3

IUPAC Name: [4-(dibenzylamino)phenyl]boronic acid
Synonyms: 4-(dibenzylamino)phenylboronic acid
Molecular Weight (g/mol): 317.2
Molecular Formula: C20H20BNO2
InChi Key: KLGQJKOCSXZOBV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results