accessibility menu, dialog, popup

UserName

CAS RN 15717-17-6

H2NSCl

CAS RN 15717-17-6

IUPAC Name: 2-chlorobenzene-1-carbothioamide
Synonyms: 2-chlorobenzenecarbothioamide
Molecular Weight (g/mol): 171.64
Molecular Formula: C7H6ClNS
InChi Key: FLQYOORLPNYQEV-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results