accessibility menu, dialog, popup

UserName

CAS RN 1547-36-0

OHOHOFFFFFF

CAS RN 1547-36-0

IUPAC Name: 4,4,4-trifluoro-3-hydroxy-3-(trifluoromethyl)butanoic acid
Synonyms: 4,4,4-trifluoro-3-hydroxy-3-(trifluoromethyl)butanoic acid
Molecular Weight (g/mol): 226.07
Molecular Formula: C5H4F6O3
InChi Key: NOHJBOWARMTILE-UHFFFAOYSA-N
SMILES: OC(=O)CC(O)(C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results