accessibility menu, dialog, popup

UserName

CAS RN 154674-81-4

CH3CH3OOOOOHNHOH3CHNOCH3H3CNHOHONHOH3C

CAS RN 154674-81-4

IUPAC Name: 5-(2,6-dimethylbenzoyloxy)-3-(2-{2-[2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido}propanamido)-4-oxopentanoic acid
Synonyms: 5-(2,6-dimethylbenzoyloxy)-3-(2-{2-[2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido}propanamido)-4-oxopentanoic acid
Molecular Weight (g/mol): 654.72
Molecular Formula: C33H42N4O10
InChi Key: LOACDHZQOPEFKK-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)C(CC1=CC=C(O)C=C1)NC(C)=O)C(=O)NC(C)C(=O)NC(CC(O)=O)C(=O)COC(=O)C1=C(C)C=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)