accessibility menu, dialog, popup

UserName

CAS RN 154590-33-7

NNHNOOF

CAS RN 154590-33-7

IUPAC Name: 1-(2-fluoro-4-nitrophenyl)piperazine
Synonyms: 1-(2-fluoro-4-nitrophenyl)piperazine
Molecular Weight (g/mol): 225.22
Molecular Formula: C10H12FN3O2
InChi Key: ZILOTWJFFLIFMZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(C=C1)N1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results