CAS RN 149949-30-4
CAS RN 149949-30-4
IUPAC Name:
2,6-difluoro-3-methoxybenzaldehyde
Synonyms:
2,6-difluoro-3-methoxybenzaldehyde
Molecular Weight (g/mol):
172.13
Molecular Formula:
C8H6F2O2
InChi Key:
WJCGNXHXQFFRKB-UHFFFAOYSA-N
SMILES:
COC1=C(F)C(C=O)=C(F)C=C1
1
–
1
of
1
results
1
2,6-Difluoro-3-methoxybenzaldehyde, 97%, Thermo Scientific™
CAS: 149949-30-4 Molecular Formula: C8H6F2O2 Molecular Weight (g/mol): 172.131 MDL Number: MFCD04115899 InChI Key: WJCGNXHXQFFRKB-UHFFFAOYSA-N Synonym: pubchem16517,acmc-1c0do,2,4-difluoro-3-formylanisole,2,6-difluoro-3-methoxy benzaldehyde,benzaldehyde,2,6-difluoro-3-methoxy,benzaldehyde, 2,6-difluoro-3-methoxy,2,6-bis fluoranyl-3-methoxy-benzaldehyde,4s,4as,5ar,12as-9-amino-4,7-bis dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-1,11-dioxo-2-naphthacenecarboxamide hydrochloride 1:1 PubChem CID: 599073 IUPAC Name: 2,6-difluoro-3-methoxybenzaldehyde SMILES: COC1=C(C(=C(C=C1)F)C=O)F