CAS RN 149391-23-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(mercaptocyclohexane)titanium tetrachloride, 99.9% (metals basis), Thermo Scientific™
CAS: 149391-23-1 MDL Number: MFCD00171702 Synonym: Tetrachlorobis(cyclohexylmercapto)titanium(IV)