accessibility menu, dialog, popup

UserName

H3CONNI

CAS RN 146697-87-2

IUPAC Name: 1-(ethoxymethyl)-2-iodo-1H-imidazole
Synonyms: 1-(ethoxymethyl)-2-iodoimidazole
Molecular Weight (g/mol): 252.06
Molecular Formula: C6H9IN2O
InChi Key: YISOPKRYYGYKJQ-UHFFFAOYSA-N
SMILES: CCOCN1C=CN=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results