CAS RN 146196-91-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Flavanone-d5, TRC
CAS: 146196-91-0 Molecular Formula: C15H7D5O2 Molecular Weight (g/mol): 229.29 Synonym: 2,3-Dihydro-2-(phenyl-d5)-4H-1-benzopyran-4-one,(+/-)-Flavanone-d5,2-(Phenyl-d5)-3,4-dihydro-2H-benzopyran-4-one,2-(Phenyl-d5)-3,4-dihydro-benzopyran-4-one,2-(Phenyl-d5)-4-chromanone,4-Flavanone-d5,NSC 50393-d5,dl-Flavanone-d5 SMILES: [2H]c1c([2H])c([2H])c(C2CC(=O)c3ccccc3O2)c([2H])c1[2H]